Thermo Scientific Chemicals

1-Bromononane, 97%, Thermo Scientific Chemicals

Catalog number: 107205000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromononane, 97%, Thermo Scientific Chemicals

Catalog number: 107205000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 107205000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
693-58-3
IUPAC Name
1-bromononane
Molecular Formula
C9H19Br
InChI Key
AYMUQTNXKPEMLM-UHFFFAOYSA-N
SMILES
CCCCCCCCCBr
Appearance (Color)
Clear colorless to golden
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4530 to 1.4550 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text