2-Bromopentane, 90%, tech., Thermo Scientific Chemicals
2-Bromopentane, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromopentane, 90%, tech., Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number 107231000
Price (USD)/ Each
160.65
Online exclusive
178.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
160.65
Online exclusive
178.00 
Save 17.35 (10%)
Add to cart
2-Bromopentane, 90%, tech., Thermo Scientific Chemicals
Catalog number107231000
Price (USD)/ Each
160.65
Online exclusive
178.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS107-81-3
IUPAC Name2-bromopentane
Molecular FormulaC5H11Br
InChI KeyLGAJYTCRJPCZRJ-UHFFFAOYNA-N
SMILESCCCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=88 % Typical
GC=<12 % (3-Bromopentane) Typical
Refractive index1.4393 to 1.4413 (20°C, 589 nm)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only