9-Bromophenanthrene, 96%, Thermo Scientific Chemicals
9-Bromophenanthrene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

9-Bromophenanthrene, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 107240100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS573-17-1
IUPAC Name9-bromophenanthrene
Molecular FormulaC14H9Br
InChI KeyRSQXKVWKJVUZDG-UHFFFAOYSA-N
SMILESBrC1=CC2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Infrared spectrumConforms
Appearance (Form)Powder
Melting point60°C to 65°C
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only