Thermo Scientific Chemicals

9-Bromophenanthrene, 96%, Thermo Scientific Chemicals

Catalog number: 107241000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

9-Bromophenanthrene, 96%, Thermo Scientific Chemicals

Catalog number: 107241000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 107241000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
573-17-1
IUPAC Name
9-bromophenanthrene
Molecular Formula
C14H9Br
InChI Key
RSQXKVWKJVUZDG-UHFFFAOYSA-N
SMILES
BrC1=CC2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
Light yellow
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
60°C to 65°C
GC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text