2-Bromopropane, 99%
2-Bromopropane, 99%
2-Bromopropane, 99%
Thermo Scientific Chemicals

2-Bromopropane, 99%

CAS: 75-26-3 | C3H7Br | 122.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 107320010
Price (USD)
164.65
Special offer
Online exclusive
Ends: 26-Sep-2025
204.00
Save 39.35 (19%)
Each
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
164.65
Special offer
Online exclusive
Ends: 26-Sep-2025
204.00
Save 39.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS75-26-3
IUPAC Name2-bromopropane
Molecular FormulaC3H7Br
InChI KeyNAMYKGVDVNBCFQ-UHFFFAOYSA-N
SMILESCC(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to light brown
Refractive index1.4250 to 1.4270 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link