2-Bromopropene, 99%, stabilized, Thermo Scientific Chemicals
2-Bromopropene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromopropene, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass bottle
Catalog number 107340500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-bromoprop-1-ene
Molecular FormulaC3H5Br
InChI KeyPHMRPWPDDRGGGF-UHFFFAOYSA-N
SMILESCC(Br)=C
Molecular Weight (g/mol)120.98
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
StabilizerApprox. 100 ppm BHT
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only