N-(3-Bromopropyl)phthalimide, 98%, Thermo Scientific Chemicals
N-(3-Bromopropyl)phthalimide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(3-Bromopropyl)phthalimide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 107391000
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
Add to cart
N-(3-Bromopropyl)phthalimide, 98%, Thermo Scientific Chemicals
Catalog number107391000
Price (USD)/ Each
101.65
Online exclusive
113.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5460-29-7
IUPAC Name2-(3-bromopropyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC11H10BrNO2
InChI KeyVKJCJJYNVIYVQR-UHFFFAOYSA-N
SMILESBrCCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point70°C to 76°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only