Thermo Scientific Chemicals

2-Bromopyridine, 99%, Thermo Scientific Chemicals

Catalog number: 107405000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromopyridine, 99%, Thermo Scientific Chemicals

Catalog number: 107405000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 107405000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
109-04-6
IUPAC Name
2-bromopyridine
Molecular Formula
C5H4BrN
InChI Key
IMRWILPUOVGIMU-UHFFFAOYSA-N
SMILES
BrC1=CC=CC=N1
Appearance (Color)
Clear colorless to yellow to light brown
GC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.5700 to 1.5730 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text