Thermo Scientific Chemicals

4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 107420050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 107420050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 107420050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
19524-06-2
IUPAC Name
4-bromopyridine hydrochloridyl
Molecular Formula
C5H4BrClN
InChI Key
PSPIMDWZDKGZBO-UHFFFAOYSA-N
SMILES
[Cl].BrC1=CC=NC=C1
Appearance (Color)
White to light yellow-pink
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text