Gibco™

1X CD CHO Medium

Catalog number: 10743001
Have Questions?
Gibco™

1X CD CHO Medium

Catalog number: 10743001
Have Questions?
Catalog Number
10743001
also known as 10743-001
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
10743002
also known as 10743-002
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
10743001
also known as 10743-001
10 L
-
10743002
also known as 10743-002
20 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
CD CHO medium is a protein-free, serum-free, chemically-defined medium optimized for the growth of Chinese hamster ovary (CHO) cells and expression of recombinant proteins in suspension culture. With a proven track record of quality for more than 10 years, CD CHO medium contains no proteins or peptide components of animal, plant, or synthetic origin, as well as no undefined lysates or hydrolysates.

When to Use CD CHO Medium
• Your GS CHO cells are recommended to be grown in CD CHO medium
• Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

When to Consider Another Gibco™ Medium

Consider CD FortiCHO™ medium when:
• The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
• Maximum batch culture cell densities and protein titers are needed
• You have time to adapt cells into a new medium in an effort to maximize titers
• Cell health at harvest is a priority for downstream processing

Consider CD OptiCHO™ medium when:
• You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
• Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
• Your CHO cell line is “finicky,” i.e. hard to grow or adapt

Consider CD DG44 medium when:
• You use parental DG44 or DXB11 CHO cells (dhfr-)

Dry Media Formulations
Gibco™ liquid media are available in an AGT™ (Advanced Granulation Technology) dry media format that is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. AGT™ media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their liquid media counterparts.

Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.

Specifications

Concentration Ratio
1 :1
Culture Environment
CO2
Cell Line
CHO
Cell Type
Ovarian Cells
Classification
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration
1 X
Culture Type
Suspension Cell Culture
Form
Liquid
Product Type
CD CHO Medium
Serum Level
Serum-free
Sterility
Sterile-filtered
With Additives
Sodium Pyruvate, Sodium Bicarbonate, Non Essential Amino Acids (NEAA)
Without Additives
No Glutamine, No Phenol Red, No Thymidine, No Hypoxanthine
Endotoxin Level
Low
Format
Universal Bag
Included Antibiotics
No antibiotics
Product Line
Gibco™
Purity or Quality Grade
Cell Culture Grade
Quantity
10 L
Shipping Condition
Room Temperature

Contents & Storage

Store at 2-8°C in the dark.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text