4-Bromoresorcinol, 98%, Thermo Scientific Chemicals
4-Bromoresorcinol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromoresorcinol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 107430250
Price (USD)/ Each
306.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
361.00 
Save 54.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
306.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
361.00 
Save 54.35 (15%)
Add to cart
4-Bromoresorcinol, 98%, Thermo Scientific Chemicals
Catalog number107430250
Price (USD)/ Each
306.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
361.00 
Save 54.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-bromobenzene-1,3-diol
Molecular FormulaC6H5BrO2
InChI KeyMPCCNXGZCOXPMG-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C(O)=C1
Molecular Weight (g/mol)189.01
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point99°C to 104°C
Appearance (Color)Beige to pink
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only