2-Bromotoluene, 99%, Thermo Scientific Chemicals
2-Bromotoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromotoluene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number 107471000
Price (USD)/ Each
169.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
199.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
169.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
199.00 
Save 29.35 (15%)
Add to cart
2-Bromotoluene, 99%, Thermo Scientific Chemicals
Catalog number107471000
Price (USD)/ Each
169.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
199.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
CAS95-46-5
IUPAC Name1-bromo-2-methylbenzene
Molecular FormulaC7H7Br
InChI KeyQSSXJPIWXQTSIX-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.5550 to 1.5570 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only