Bromotrichloromethane, 99%, Thermo Scientific Chemicals
Bromotrichloromethane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromotrichloromethane, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 107491000
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
82.60 
Save 11.95 (14%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
82.60 
Save 11.95 (14%)
Add to cart
Bromotrichloromethane, 99%, Thermo Scientific Chemicals
Catalog number107491000
Price (USD)/ Each
70.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
82.60 
Save 11.95 (14%)
-
Add to cart
Chemical Identifiers
CAS75-62-7
IUPAC Namebromotrichloromethane
Molecular FormulaCBrCl3
InChI KeyXNNQFQFUQLJSQT-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5055 to 1.5075 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only