Thermo Scientific Chemicals

5-Bromo-DL-tryptophan, 99%, Thermo Scientific Chemicals

Catalog number: 107510010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

5-Bromo-DL-tryptophan, 99%, Thermo Scientific Chemicals

Catalog number: 107510010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 107510010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6548-09-0
IUPAC Name
2-amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
Molecular Formula
C11H11BrN2O2
InChI Key
KZDNJQUJBMDHJW-UHFFFAOYSA-N
SMILES
NC(CC1=CNC2=CC=C(Br)C=C12)C(O)=O
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystalline powder
HPLC
>=98.5 %
Appearance (Color)
White to light beige to light grey
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text