Thermo Scientific Chemicals

Butadiene sulfone, 98%, Thermo Scientific Chemicals

Catalog number: 107610025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Butadiene sulfone, 98%, Thermo Scientific Chemicals

Catalog number: 107610025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 107610025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77-79-2
IUPAC Name
2,5-dihydro-1λ⁶-thiophene-1,1-dione
Molecular Formula
C4H6O2S
InChI Key
MBDNRNMVTZADMQ-UHFFFAOYSA-N
SMILES
O=S1(=O)CC=CC1
Appearance (Form)
Adhering powder or crystals and/or chunks
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
63°C to 67°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text