Thermo Scientific Chemicals

(±)-1,3-Butanediol, 99%, Extra Pure, Thermo Scientific Chemicals

Catalog number: 107620010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

(±)-1,3-Butanediol, 99%, Extra Pure, Thermo Scientific Chemicals

Catalog number: 107620010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 107620010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-88-0
IUPAC Name
butane-1,3-diol
Molecular Formula
C4H10O2
InChI Key
PUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILES
CC(O)CCO
Color scale
=<10 APHA
Refractive index
1.4390 to 1.4410 (20°C, 589 nm)
Water
=<0.5 %
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text