(±)-1,3-Butanediol, 99%, extra pure
(±)-1,3-Butanediol, 99%, extra pure
(±)-1,3-Butanediol, 99%, extra pure
(±)-1,3-Butanediol, 99%, extra pure
Thermo Scientific Chemicals

(±)-1,3-Butanediol, 99%, extra pure

CAS: 107-88-0 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 107622500
Price (USD)
28.65
Special offer
Online exclusive
Ends: 26-Sep-2025
35.70
Save 7.05 (20%)
Each
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 26-Sep-2025
35.70
Save 7.05 (20%)
Each
Add to cart
Chemical Identifiers
CAS107-88-0
IUPAC Namebutane-1,3-diol
Molecular FormulaC4H10O2
InChI KeyPUPZLCDOIYMWBV-UHFFFAOYNA-N
SMILESCC(O)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
Water=<0.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link