Thermo Scientific Chemicals

1,4-Butane sultone, 99+%, Thermo Scientific Chemicals

Catalog number: 107670250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,4-Butane sultone, 99+%, Thermo Scientific Chemicals

Catalog number: 107670250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 107670250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1633-83-6
Water
=<0.1 % (K.F.)
Titration with TBAH
=<0.1 % 4-Hydoxybutanesulfonic acid
Appearance (Form)
Liquid
GC
>=99.0 %
Refractive index
1.4600 to 1.4640 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text