1-Butanol, 99%, extra pure, Thermo Scientific Chemicals
1-Butanol, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Butanol, 99%, extra pure, Thermo Scientific Chemicals

1-Butanol, 99%, C4H10O, CAS Number-71-36-3, 1-butanol, 1-hydroxybutane, butanol, butyl alcohol, butyl hydroxide, methylolpropane, n-butanol, n-butyl alcohol, propylcarbinol, propylmethanol, 25L, Vapor Pressure: 6.7mbar at 20 deg.C, 01, 367, 117.6 deg.C, 100.0, 98.5, CHEBI:28885, Undesignated
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 107690010
Price (USD)/ Each
45.65
Online exclusive
53.00 
Save 7.35 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
53.00 
Save 7.35 (14%)
Add to cart
1-Butanol, 99%, extra pure, Thermo Scientific Chemicals
Catalog number107690010
Price (USD)/ Each
45.65
Online exclusive
53.00 
Save 7.35 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
Molecular Weight (g/mol)74.12
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.2 % (Coulometric)
GC>=98.5 %
Refractive index1.3980 to 1.4000 (20°C, 589 nm)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only