tert-Butanol, 99.5%, extra pure, Thermo Scientific Chemicals
tert-Butanol, 99.5%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butanol, 99.5%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Plastic Bottle
Catalog number 107710025
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
Add to cart
tert-Butanol, 99.5%, extra pure, Thermo Scientific Chemicals
Catalog number107710025
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.3860 to 1.3880 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.03 % (K.F.)
Appearance (Form)Liquid after melting
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only