cis-2-Butene-1,4-diol, 97%, Thermo Scientific Chemicals
cis-2-Butene-1,4-diol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-2-Butene-1,4-diol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 107720025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2Z)-but-2-ene-1,4-diol
Molecular FormulaC4H8O2
InChI KeyORTVZLZNOYNASJ-UPHRSURJSA-N
SMILESOC\C=C/CO
Molecular Weight (g/mol)88.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 % (cis-isomer)
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4770 to 1.4800 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only