Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
10777019 | 5000 units |
The robust molecular structure of the RNaseOUT™ inhibitor together with a very high binding affinity for pancreatic-type RNases ensures superior protection of RNA from degradation making RNaseOUT™ highly effective and reliable in various molecular biology applications.
Features
Applications
Source
Purified by affinity chromatography from E. coli expressing a cloned porcine gene.
Performance and quality testing
Unit definition
One unit inhibits 5 ng of RNase A by 50% using cytidine 2', 3' cyclic monophosphate (cCMP) as the substrate.
Unit reaction conditions
100 mM Tris-acetate (pH 6.5), 1 mM EDTA, 0.2 mM cCMP, 2 μg RNase A in 1 mL from 0 to 10 min at 25°C
Share catalog number, name or link