tert-Butylacetic Acid, 98%, Thermo Scientific Chemicals
tert-Butylacetic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butylacetic Acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
250 mL
Packaging:
Glass bottle
Catalog number 107770500
Price (USD)/ Each
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1070-83-3
IUPAC Name3,3-dimethylbutanoic acid
Molecular FormulaC6H12O2
InChI KeyMLMQPDHYNJCQAO-UHFFFAOYSA-N
SMILESCC(C)(C)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration with NaOH>=97.5 %
Infrared spectrumConforms
Refractive index1.4090 to 1.4110 (20°C, 589 nm)
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only