Thermo Scientific Chemicals

tert-Butylacetic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 107772500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

tert-Butylacetic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 107772500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 107772500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1070-83-3
IUPAC Name
3,3-dimethylbutanoic acid
Molecular Formula
C6H12O2
InChI Key
MLMQPDHYNJCQAO-UHFFFAOYSA-N
SMILES
CC(C)(C)CC(O)=O
Appearance (Form)
Liquid
Titration with NaOH
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.4090 to 1.4110 (20°C, 589 nm)
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text