Thermo Scientific Chemicals

sec-Butylbenzene, 99+%, Thermo Scientific Chemicals

Catalog number: 107862500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

sec-Butylbenzene, 99+%, Thermo Scientific Chemicals

Catalog number: 107862500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 107862500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
135-98-8
IUPAC Name
(butan-2-yl)benzene
Molecular Formula
C10H14
InChI Key
ZJMWRROPUADPEA-UHFFFAOYNA-N
SMILES
CCC(C)C1=CC=CC=C1
Refractive index
1.4890 to 1.4910 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=99.0 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text