Thermo Scientific Chemicals

tert-Butyl carbazate, 99%, Thermo Scientific Chemicals

Catalog number: 107880250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

tert-Butyl carbazate, 99%, Thermo Scientific Chemicals

Catalog number: 107880250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 107880250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
870-46-2
IUPAC Name
(tert-butoxy)carbohydrazide
Molecular Formula
C5H12N2O2
InChI Key
DKACXUFSLUYRFU-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NN
Appearance (Form)
Crystals or fused solid
Titration with HClO4
98.5 to 101.5 %
Appearance (Color)
White to light yellow to light beige
Infrared spectrum
Conforms
Melting point
37°C to 41°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text