Thermo Scientific Chemicals

4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 107900050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 107900050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 107900050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
98-52-2
IUPAC Name
4-tert-butylcyclohexan-1-ol
Molecular Formula
C10H20O
InChI Key
CCOQPGVQAWPUPE-UHFFFAOYSA-N
SMILES
CC(C)(C)C1CCC(O)CC1
Melting point
60°C to 69°C (depending on cis/trans ratio)
Appearance (Color)
White
Infrared spectrum
Conforms
GC
>=98.5 %
Ratio
Cis/trans ratio: approximately 70% trans, 30% cis

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text