Thermo Scientific Chemicals

2-tert-Butyl-4-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 107961000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-tert-Butyl-4-methylphenol, 99%, Thermo Scientific Chemicals

Catalog number: 107961000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 107961000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2409-55-4
IUPAC Name
2-tert-butyl-4-methylphenol
Molecular Formula
C11H16O
InChI Key
IKEHOXWJQXIQAG-UHFFFAOYSA-N
SMILES
CC1=CC=C(O)C(=C1)C(C)(C)C
Appearance (Color)
White to yellow to light beige
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Crystalline mass
Melting point
50°C to 53°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text