Butyraldehyde, 99%
Butyraldehyde, 99%
Butyraldehyde, 99%
Thermo Scientific Chemicals

Butyraldehyde, 99%

CAS: 123-72-8 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 108091000
Price (USD)
40.65
Special offer
Online exclusive
Ends: 26-Sep-2025
49.60
Save 8.95 (18%)
Each
In stock
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 26-Sep-2025
49.60
Save 8.95 (18%)
Each
Add to cart
Chemical Identifiers
CAS123-72-8
IUPAC Namebutanal
Molecular FormulaC4H8O
InChI KeyZTQSAGDEMFDKMZ-UHFFFAOYSA-N
SMILESCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3770 to 1.3830 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link