Invitrogen™

Second Strand Buffer

Catalog number: 10812014
Have Questions?
Invitrogen™

Second Strand Buffer

Catalog number: 10812014
Have Questions?
Catalog Number
10812014
Unit Size
0.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
108120140.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Second Strand Buffer is used with SuperScript™ cDNA Synthesis Systems to generate second-strand cDNA from mRNA in the first-strand reaction. It is supplied as a 5X concentrate and should be diluted 1:5 (1 part 5X Second Strand Buffer + 4 parts other components) prior to use.

Quality control
This product has passed quality control assays that verify the absence of detectable levels of ribonuclease activity.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
5X
Shipping Condition
Approved for shipment on Wet or Dry Ice

Contents & Storage

Second-Strand Buffer is supplied at a 5X concentration. Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text