Thermo Scientific Chemicals

n-Butyric Anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 108121000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

n-Butyric Anhydride, 98%, Thermo Scientific Chemicals

Catalog number: 108121000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 108121000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-31-0
IUPAC Name
butanoyl butanoate
Molecular Formula
C8H14O3
InChI Key
YHASWHZGWUONAO-UHFFFAOYSA-N
SMILES
CCCC(=O)OC(=O)CCC
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
GC
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.4120 to 1.4140 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text