Thermo Scientific Chemicals

S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals

Catalog number: 108151000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

S-Butyrylthiocholine iodide, 98%, Thermo Scientific Chemicals

Catalog number: 108151000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 108151000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1866-16-6
IUPAC Name
[2-(butanoylsulfanyl)ethyl]trimethylazanium iodide
Molecular Formula
C9H20INOS
InChI Key
WEQAAFZDJROSBF-UHFFFAOYSA-M
SMILES
[I-].CCCC(=O)SCC[N+](C)(C)C
Appearance (Form)
Crystalline powder or crystals
Melting point
172°C to 176°C
Appearance (Color)
White
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text