BlueJuice Gel Loading Buffer (10X) is designed for easy loading and tracking of DNA samples in agarose or native polyacrylamideRead more
Have Questions?
Catalog Number
Quantity
10816015
3 x 1 mL
Catalog number 10816015
Price (USD)
73.65
Online Exclusive
78.25
Save 4.60 (6%)
Each
Add to cart
Quantity:
3 x 1 mL
Request bulk or custom format
Price (USD)
73.65
Online Exclusive
78.25
Save 4.60 (6%)
Each
Add to cart
BlueJuice Gel Loading Buffer (10X) is designed for easy loading and tracking of DNA samples in agarose or native polyacrylamide gels. Visualization of DNA bands will not be obscured by the tracking dyes because they run outside the limits of most DNA samples.
Product use The recommended concentration of this buffer for use with all DNA samples run on agarose gels is 1X. For acrylamide gels, the recommended concentration is 0.5 X.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration10X
Gel CompatibilityAgarose Gels
Product LineBlueJuice™
Product TypeGel Loading Buffer
Quantity3 x 1 mL
Sample Loading Volume1 mL
Shelf Life6 Months
Shipping ConditionRoom Temperature
Gel TypeAgarose
Label or DyeBromophenol Blue
Unit SizeEach
Contents & Storage
• 3 x 1 mL 10X BlueJuice Gel Loading Buffer
Store at room temperature or at 4°C for up to 6 months. For longer periods, store at -20°C.