Calmagite, Indicator Grade, Pure
Calmagite, Indicator Grade, Pure
Thermo Scientific Chemicals

Calmagite, Indicator Grade, Pure

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 108170100
Price (USD)/ Each
64.65
Online exclusive
71.30 
Save 6.65 (9%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
71.30 
Save 6.65 (9%)
Add to cart
Calmagite, Indicator Grade, Pure
Catalog number108170100
Price (USD)/ Each
64.65
Online exclusive
71.30 
Save 6.65 (9%)
-
Add to cart
Chemical Identifiers
CAS3147-14-6
IUPAC Name4-[2-(2-hydroxy-5-methylphenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonic acid
Molecular FormulaC17H14N2O5S
InChI KeyASFVMSDYPYMUFL-UHFFFAOYSA-N
SMILESCC1=CC(NN=C2C(=O)C=C(C3=CC=CC=C23)S(O)(=O)=O)=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown to black
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Specific extinction E (1%/1cm)>=300 (610 nm, in buffer)
Suitability(for Ca + Mg determination) Passes test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only