Thermo Scientific Chemicals

D(+)-Camphoric acid, 99%, Thermo Scientific Chemicals

Catalog number: 108205000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

D(+)-Camphoric acid, 99%, Thermo Scientific Chemicals

Catalog number: 108205000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 108205000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
124-83-4
IUPAC Name
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate
Molecular Formula
C10H14O4
InChI Key
LSPHULWDVZXLIL-LDWIPMOCSA-L
SMILES
CC1(C)[C@H](CC[C@@]1(C)C([O-])=O)C([O-])=O
Infrared spectrum
Conforms
Titration with NaOH
>=98.5 %
Appearance (Form)
Crystalline powder
Melting point
183°C to 187°C
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text