Thermo Scientific Chemicals

ε-Caprolactam, 99+%, Thermo Scientific Chemicals

Catalog number: 108230025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

ε-Caprolactam, 99+%, Thermo Scientific Chemicals

Catalog number: 108230025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 108230025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105-60-2
IUPAC Name
azepan-2-one
Molecular Formula
C6H11NO
InChI Key
JBKVHLHDHHXQEQ-UHFFFAOYSA-N
SMILES
O=C1CCCCCN1
Appearance (Form)
Crystals or flakes
Melting point
68°C to 71°C
Infrared spectrum
Conforms
GC
>=99.0 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text