(Carbethoxymethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals
(Carbethoxymethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Carbethoxymethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 108295000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameethyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular FormulaC22H21O2P
InChI KeyIIHPVYJPDKJYOU-UHFFFAOYSA-N
SMILESCCOC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)348.38
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Melting point124°C to 129°C
Infrared spectrumConforms
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only