Thermo Scientific Chemicals

3-Carbethoxy-4-piperidone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 108330050
5 g, Each
Thermo Scientific Chemicals

3-Carbethoxy-4-piperidone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 108330050
5 g, Each
Quantity
Catalog number: 108330050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4644-61-5
IUPAC Name
ethyl 4-oxopiperidine-3-carboxylate hydrochloridyl
Molecular Formula
C8H13ClNO3
InChI Key
OTNOTUGMYUCNQB-UHFFFAOYNA-N
SMILES
[Cl].CCOC(=O)C1CNCCC1=O
Melting point
169°C to 175°C
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text