Thermo Scientific Chemicals

DL-Carnitine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 108451000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

DL-Carnitine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 108451000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 108451000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
461-05-2
IUPAC Name
(3-carboxy-2-hydroxypropyl)trimethylazanium chloride
Molecular Formula
C7H16ClNO3
InChI Key
JXXCENBLGFBQJM-UHFFFAOYNA-N
SMILES
[Cl-].C[N+](C)(C)CC(O)CC(O)=O
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Melting point
194°C to 200°C
Titration with HClO4
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text