D(+)-Cellobiose, 98%
D(+)-Cellobiose, 98%
Thermo Scientific Chemicals

D(+)-Cellobiose, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 108460050
Price (USD)/ Each
28.10
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
28.10
Add to cart
D(+)-Cellobiose, 98%
Catalog number108460050
Price (USD)/ Each
28.10
-
Add to cart
Chemical Identifiers
CAS528-50-7
IUPAC Name2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+33° to +35° (20°C, 589 nm) (c=4, water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only