Thermo Scientific Chemicals

Chelidamic acid monohydrate, 98%

Catalog number: 108470050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Chelidamic acid monohydrate, 98%

Catalog number: 108470050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 108470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
199926-39-1
IUPAC Name
4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid hydrate
Molecular Formula
C7H7NO6
InChI Key
SNGPHFVJWBKEDG-UHFFFAOYSA-N
SMILES
O.OC(=O)C1=CC(=O)C=C(N1)C(O)=O
Appearance (Form)
Crystalline powder
Water
=<10 %
Titration with NaOH
>=97.5 %
Appearance (Color)
White to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text