Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 108502500
Price (USD)/ Each
39.10
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
39.10
Add to cart
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Catalog number108502500
Price (USD)/ Each
39.10
-
Add to cart
Chemical Identifiers
CAS97-97-2
IUPAC Name2-chloro-1,1-dimethoxyethane
Molecular FormulaC4H9ClO2
InChI KeyCRZJPEIBPQWDGJ-UHFFFAOYSA-N
SMILESCOC(CCl)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.4140 to 1.4160 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only