Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals
Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
4 kg
Packaging:
Plastic bottle
Catalog number 108510010
Price (USD)/ Each
55.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
65.60 
Save 9.95 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
55.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
65.60 
Save 9.95 (15%)
Add to cart
Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals
Catalog number108510010
Price (USD)/ Each
55.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
65.60 
Save 9.95 (15%)
-
Add to cart
Chemical Identifiers
CAS79-11-8
IUPAC Name2-chloroacetic acid
Molecular FormulaC2H3ClO2
InChI KeyFOCAUTSVDIKZOP-UHFFFAOYSA-N
SMILESOC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline flakes
Titration with NaOH>=98.5 %
Appearance (Color)White
Infrared spectrumConforms
Melting point61°C to 63°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only