Thermo Scientific Chemicals

Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals

Catalog number: 108512500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Chloroacetic acid, 99%, flakes, Thermo Scientific Chemicals

Catalog number: 108512500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 108512500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-11-8
IUPAC Name
2-chloroacetic acid
Molecular Formula
C2H3ClO2
InChI Key
FOCAUTSVDIKZOP-UHFFFAOYSA-N
SMILES
OC(=O)CCl
Appearance (Form)
Crystalline flakes
Titration with NaOH
>=98.5 %
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
61°C to 63°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text