4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
Packaging:
Glass bottle
Catalog number 108540010
Price (USD)/ Each
179.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
211.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
179.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
211.00 
Save 31.35 (15%)
Add to cart
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
Catalog number108540010
Price (USD)/ Each
179.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
211.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-(4-chlorophenyl)ethan-1-one
Molecular FormulaC8H7ClO
InChI KeyBUZYGTVTZYSBCU-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol)154.59
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5539 to 1.5559 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid after melting
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only