trans-3-chloroacrylic acid, 99%, Thermo Scientific Chemicals
trans-3-chloroacrylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-3-chloroacrylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 108550010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2345-61-1
IUPAC Name(2E)-3-chloroprop-2-enoic acid
Molecular FormulaC3H3ClO2
InChI KeyMHMUCYJKZUZMNJ-OWOJBTEDSA-N
SMILESOC(=O)\C=C\Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point80°C to 86°C
Appearance (Form)Powder or crystals
Appearance (Color)White to beige to grey
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only