2-Chloroacrylonitrile, 99%, stabilized, Thermo Scientific Chemicals
2-Chloroacrylonitrile, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroacrylonitrile, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 108560050
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.10 
Save 7.45 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.10 
Save 7.45 (14%)
Add to cart
2-Chloroacrylonitrile, 99%, stabilized, Thermo Scientific Chemicals
Catalog number108560050
Price (USD)/ Each
44.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
52.10 
Save 7.45 (14%)
-
Add to cart
Chemical Identifiers
CAS920-37-6
IUPAC Name2-chloroprop-2-enenitrile
Molecular FormulaC3H2ClN
InChI KeyOYUNTGBISCIYPW-UHFFFAOYSA-N
SMILESClC(=C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4290 to 1.4310 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only