4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloroaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 108590010
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
Add to cart
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Catalog number108590010
Price (USD)/ Each
98.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
116.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-chloroaniline
Molecular FormulaC6H6ClN
InChI KeyQSNSCYSYFYORTR-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1
Molecular Weight (g/mol)127.57
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to brown to purple
Melting point68°C to 72°C
AppearanceMay darken on storage
GC>=97.5 %
Appearance (Form)Crystalline solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only