Thermo Scientific Chemicals

4-Chloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 108595000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Chloroaniline, 98%, Thermo Scientific Chemicals

Catalog number: 108595000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 108595000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-47-8
IUPAC Name
4-chloroaniline
Molecular Formula
C6H6ClN
InChI Key
QSNSCYSYFYORTR-UHFFFAOYSA-N
SMILES
NC1=CC=C(Cl)C=C1
Appearance (Color)
Beige to brown to purple
Melting point
68°C to 72°C
Appearance
May darken on storage
GC
>=97.5 %
Appearance (Form)
Crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text