Thermo Scientific Chemicals

1-(4-Chlorobenzhydryl)piperazine, 90-95%, Thermo Scientific Chemicals

Catalog number: 108650050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

1-(4-Chlorobenzhydryl)piperazine, 90-95%, Thermo Scientific Chemicals

Catalog number: 108650050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 108650050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
303-26-4
IUPAC Name
hydrogen 1-[(4-chlorophenyl)(phenyl)methyl]piperazine chloride
Molecular Formula
C17H20Cl2N2
InChI Key
XFIUZNUONMJUIH-UHFFFAOYNA-N
SMILES
[H+].[Cl-].ClC1=CC=C(C=C1)C(N1CCNCC1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Titration with HClO4
>=90.0 % (On dry substance)
TLC
>=90 %
Appearance (Color)
White to light beige
Melting point
65°C to 75°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text